KEY466133355 Key Organics / BIONET

1-(4-bromo-2-methyl-6-nitrophenyl)-1H-pyrrole

  • MDL number MFCD03617774
  • Cas Number : 860650-88-0
  • Molecular Weight : 281.109

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7066

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0850_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart