KEY466133358 Key Organics / BIONET

3-(quinolin-2-yl)phenol

  • MDL number MFCD03617606
  • Cas Number : 87741-93-3
  • Molecular Weight : 221.259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7073

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0859_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart