KEY466133359 Key Organics / BIONET

2-(4-chlorophenoxy)-1-[1-(4-methyl-5,6,7,8-tetrahydroquinazolin-2-yl)-1H-1,2,3-triazol-4-yl]propan-1-one

  • MDL number MFCD03787483
  • Cas Number : 860650-93-7
  • Molecular Weight : 397.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7074

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0861_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart