KEY466133360 Key Organics / BIONET

(1E,4E)-1,5-bis(2-chloro-6-fluorophenyl)penta-1,4-dien-3-one

  • MDL number MFCD00121150
  • Cas Number : 346670-60-8
  • Molecular Weight : 339.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7075

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0862_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart