KEY466133374 Key Organics / BIONET

1-tert-butyl-4-{2-[(3-methylphenyl)methoxy]benzoyl}-1H-pyrazole

  • MDL number MFCD04124377
  • Cas Number : 955975-74-3
  • Molecular Weight : 348.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7142

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0276_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart