KEY466133378 Key Organics / BIONET

7-(pyrrolidine-1-sulfonyl)-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxalin-4-one

  • MDL number MFCD04124391
  • Cas Number : 1164124-56-4
  • Molecular Weight : 321.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7158

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0303_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart