KEY466133382 Key Organics / BIONET

ethyl 2-{2-[(4-hydroxy-1,3-thiazol-2-yl)methyl]-1,3-thiazol-4-yl}acetate

  • MDL number MFCD04124398
  • Cas Number : 860651-37-2
  • Molecular Weight : 284.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7166

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0318_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart