KEY466133384 Key Organics / BIONET

2-(1,2-benzoxazol-3-yl)-N-[(ethylcarbamothioyl)amino]acetamide

  • MDL number MFCD04124401
  • Cas Number : 860651-40-7
  • Molecular Weight : 278.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7169

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0321_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart