KEY466133387 Key Organics / BIONET

2-{[4-(2-fluorophenyl)-1,3-thiazol-2-yl]methyl}-1,3-thiazol-4-ol

  • MDL number MFCD04124408
  • Cas Number : 860651-46-3
  • Molecular Weight : 292.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7177

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0331_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart