KEY466133390 Key Organics / BIONET

1-{2-[(4-hydroxy-1,3-thiazol-2-yl)methyl]-4-methyl-1,3-thiazol-5-yl}ethan-1-one

  • MDL number MFCD04124418
  • Cas Number : 860651-54-3
  • Molecular Weight : 254.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7187

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0353_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart