KEY466133393 Key Organics / BIONET

2-(1,2-benzoxazol-3-yl)-N-{[(4-chlorophenyl)carbamothioyl]amino}acetamide

  • MDL number MFCD04124421
  • Cas Number : 860651-57-6
  • Molecular Weight : 360.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7191

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0357_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart