KEY466133396 Key Organics / BIONET

ethyl 2-{[4-(trifluoromethyl)phenyl]amino}-1,3-thiazole-4-carboxylate

  • MDL number MFCD04124427
  • Cas Number : 860783-84-2
  • Molecular Weight : 316.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7202

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0804_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart