KEY466133397 Key Organics / BIONET

(3Z)-3-[(4-chlorophenyl)methyl]-4-{[2-(5-methoxy-1H-indol-3-yl)ethyl]amino}pent-3-en-2-one

  • MDL number MFCD04124428
  • Cas Number : 860783-85-3
  • Molecular Weight : 396.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7203

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0805_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart