KEY466133398 Key Organics / BIONET

methyl 2-{2-amino-[1,2,4]triazolo[1,5-a]pyrimidin-7-yl}octanoate

  • MDL number MFCD04124438
  • Cas Number : 860783-94-4
  • Molecular Weight : 291.355

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7213

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart