KEY466133403 Key Organics / BIONET

1-[(2-chloro-1,3-thiazol-5-yl)methyl]piperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD04124453
  • Cas Number : 1052404-18-8
  • Molecular Weight : 297.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7233

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0848_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart