KEY466133406 Key Organics / BIONET

methyl 3-nitro-2-[(4-phenylpiperazin-1-yl)methyl]benzoate

  • MDL number MFCD04124462
  • Cas Number : 860784-17-4
  • Molecular Weight : 355.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7244

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0862_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart