KEY466133409 Key Organics / BIONET

methyl 1-methyl-4-{[(3Z)-2-oxo-2,3-dihydro-1H-indol-3-ylidene]methyl}-1H-pyrrole-2-carboxylate

  • MDL number MFCD01764881
  • Cas Number : 260781-65-5
  • Molecular Weight : 282.299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7256

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11X-0058_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart