KEY466133416 Key Organics / BIONET

2-{[2-(piperidin-1-yl)-1,3-thiazol-5-yl]methylidene}-2,3-dihydro-1H-indene-1,3-dione

  • MDL number MFCD05975037
  • Cas Number : 860784-45-8
  • Molecular Weight : 324.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7307

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11X-0828_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart