KEY466133418 Key Organics / BIONET

methyl 2-{4-[(6-chloropyridin-3-yl)methoxy]phenyl}acetate

  • MDL number MFCD05975038
  • Cas Number : 860784-47-0
  • Molecular Weight : 291.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7311

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11X-0833_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart