KEY466133419 Key Organics / BIONET

3-(2,4-difluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

  • MDL number MFCD05975039
  • Cas Number : 860784-48-1
  • Molecular Weight : 317.292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7312

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11X-0834_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart