KEY466133423 Key Organics / BIONET

3-acetyl-4-hydroxy-1-phenyl-1,2-dihydroquinolin-2-one

  • MDL number MFCD00179942
  • Cas Number : 54289-77-9
  • Molecular Weight : 279.295

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7328

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11X-0928_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart