KEY466133434 Key Organics / BIONET

N,3-bis(3-fluorophenyl)-5-(methylsulfanyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-amine

  • MDL number MFCD28016167
  • Cas Number : 1795300-32-1
  • Molecular Weight : 370.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202 - 203
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432016

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11X-9005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart