KEY466133436 Key Organics / BIONET

4-hydroxy-6-methyl-3-[(2E)-3-phenylprop-2-enoyl]-2H-pyran-2-one

  • MDL number MFCD00157334
  • Cas Number : 593278-92-3
  • Molecular Weight : 256.257

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7363

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11Y-0823_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart