KEY466133442 Key Organics / BIONET

1-(5-{1,4-dioxa-8-azaspiro[4.5]decan-8-yl}-2-nitrophenyl)ethan-1-one

  • MDL number MFCD08064701
  • Cas Number : 923106-13-2
  • Molecular Weight : 306.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7400

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11Z-0849_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart