KEY466133450 Key Organics / BIONET

3-oxo-N-phenylpyrazolidine-1-carboxamide

  • MDL number MFCD00127152
  • Cas Number : 303994-72-1
  • Molecular Weight : 205.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7418

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12A-011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart