KEY466133457 Key Organics / BIONET

(2E)-2-cyano-3-(pyridin-4-yl)prop-2-enamide

  • MDL number MFCD00215244
  • Cas Number : 40029-37-6
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7428

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12A-023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart