KEY466133485 Key Organics / BIONET

3-methoxy-3-methyl-1-[5-oxo-2-(trifluoromethyl)-5H-chromeno[2,3-b]pyridin-3-yl]urea

  • MDL number MFCD00138657
  • Cas Number : 303995-28-0
  • Molecular Weight : 367.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7505

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12B-034_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart