KEY466133486 Key Organics / BIONET

methyl 1-oxo-2-[(pyridin-2-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate

  • MDL number MFCD00140339
  • Cas Number : 303995-32-6
  • Molecular Weight : 294.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7509

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12B-043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart