KEY466133488 Key Organics / BIONET

[(E)-2-(1,3-dimethyl-4-nitro-1H-pyrazol-5-yl)ethenyl]dimethylamine

  • MDL number MFCD00244628
  • Cas Number : 303995-41-7
  • Molecular Weight : 210.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7518

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12B-054_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart