KEY466133489 Key Organics / BIONET

5-[(E)-2-[4-(diethoxymethyl)phenyl]ethenyl]-3-methyl-4-nitro-1,2-oxazole

  • MDL number MFCD00127196
  • Cas Number : 303995-43-9
  • Molecular Weight : 332.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7519

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12B-061_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart