KEY466133495 Key Organics / BIONET

4-{[(2,4-dichlorophenyl)amino]methyl}-1,2-dihydrophthalazin-1-one

  • MDL number MFCD00139367
  • Cas Number : 303995-64-4
  • Molecular Weight : 320.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7542

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12B-095_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart