KEY466133496 Key Organics / BIONET

(1Z,4E)-5-[4-(diethoxymethyl)phenyl]-1-(dimethylamino)penta-1,4-dien-3-one

  • MDL number MFCD00138870
  • Cas Number : 303995-68-8
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7549

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12B-105_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart