KEY466133506 Key Organics / BIONET

(3E)-1-{[1,1'-biphenyl]-4-yl}-3-(methoxyimino)propan-1-one

  • MDL number MFCD00127210
  • Cas Number : 303995-84-8
  • Molecular Weight : 253.301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7568

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12C-010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart