KEY466133508 Key Organics / BIONET

1-(4-benzoyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

  • MDL number MFCD00141840
  • Cas Number : 303995-88-2
  • Molecular Weight : 316.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7575

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12C-044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart