KEY466133517 Key Organics / BIONET

(E)-N-[(4-{[(4-chlorophenyl)methyl]sulfanyl}-3-nitrophenyl)methylidene]hydroxylamine

  • MDL number MFCD00139286
  • Cas Number : 303996-06-7
  • Molecular Weight : 322.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7596

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12D-005S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart