KEY466133518 Key Organics / BIONET

(E)-[(4-{[(4-chlorophenyl)methyl]sulfanyl}-3-nitrophenyl)methylidene](methoxy)amine

  • MDL number MFCD00139287
  • Cas Number : 303996-07-8
  • Molecular Weight : 336.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7597

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12D-006S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart