KEY466133521 Key Organics / BIONET

ethyl 2-{5H,6H-imidazo[2,1-b][1,3]thiazol-3-yl}acetate hydrochloride

  • MDL number MFCD00828716
  • Cas Number : 76629-19-1
  • Molecular Weight : 248.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7611

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12D-024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart