KEY466133530 Key Organics / BIONET

6-[(4-methylphenyl)sulfanyl]imidazo[2,1-b][1,3]thiazole-5-carbonitrile

  • MDL number MFCD00139733
  • Cas Number : 303996-48-7
  • Molecular Weight : 271.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7647

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12D-079S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart