KEY466133550 Key Organics / BIONET

(3E)-4-(4-methylphenyl)-1-phenylbut-3-en-2-one

  • MDL number MFCD00141308
  • Cas Number : 38661-85-7
  • Molecular Weight : 236.314

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7743

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12E-911_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart