KEY466133551 Key Organics / BIONET

ethyl 5-amino-3-{2-[3-(trifluoromethyl)benzoyloxy]ethoxy}-1,2-oxazole-4-carboxylate

  • MDL number MFCD00172370
  • Cas Number : 303997-10-6
  • Molecular Weight : 388.299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7744

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12E-913_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart