KEY466133555 Key Organics / BIONET

ethyl N-[1-(2-hydroxyethyl)-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonyl]carbamate

  • MDL number MFCD00172927
  • Cas Number : 303997-18-4
  • Molecular Weight : 271.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7759

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12E-929_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart