KEY466133560 Key Organics / BIONET

2-(dimethylamino)-6-oxo-4-(piperidin-1-yl)-6H-1,3-oxazine-5-carbonitrile

  • MDL number MFCD00171939
  • Cas Number : 303997-25-3
  • Molecular Weight : 248.286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7766

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12E-937_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart