KEY466133561 Key Organics / BIONET

4-[(2,4-difluorophenyl)amino]-2-(dimethylamino)-6-oxo-6H-1,3-oxazine-5-carbonitrile

  • MDL number MFCD00171940
  • Cas Number : 303997-26-4
  • Molecular Weight : 292.246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7767

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12E-938_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart