KEY466133568 Key Organics / BIONET

2-(4-chlorophenyl)-2-[6-(dimethylamino)pyridazin-3-yl]acetonitrile

  • MDL number MFCD00243406
  • Cas Number : 303997-51-5
  • Molecular Weight : 272.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7795

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12F-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart