KEY466133570 Key Organics / BIONET

methyl 3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-methyl-1,2-oxazole-4-carboxylate

  • MDL number MFCD00975526
  • Cas Number : 303997-60-6
  • Molecular Weight : 320.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7803

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12F-028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart