KEY466133571 Key Organics / BIONET

1-[4-(2-bromoacetyl)-1-methyl-1H-pyrrol-2-yl]-2,2,2-trichloroethan-1-one

  • MDL number MFCD00173438
  • Cas Number : 303997-66-2
  • Molecular Weight : 347.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7808

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12F-043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart