KEY466133579 Key Organics / BIONET

5-chloro-1-[(4-chlorophenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00141722
  • Cas Number : 87423-59-4
  • Molecular Weight : 306.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7890

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12F-374S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart