KEY466133580 Key Organics / BIONET

ethyl 2-{[(1E)-2-acetyl-2-[(1,3-thiazol-2-yl)carbamoyl]eth-1-en-1-yl]amino}acetate

  • MDL number MFCD00172743
  • Cas Number : 303998-68-7
  • Molecular Weight : 297.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7924

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12F-900_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart