KEY466133600 Key Organics / BIONET

methyl 3-amino-1-cyano-4-fluoro-1-(thiophen-2-yl)-1H-indene-2-carboxylate

  • MDL number MFCD00232240
  • Cas Number : 306976-63-6
  • Molecular Weight : 314.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7995

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12G-021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart