KEY466133610 Key Organics / BIONET

ethyl 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD00173065
  • Cas Number : 91396-27-9
  • Molecular Weight : 265.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8066

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12G-368S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart